Bentsoyyliperoksidi, humidified with ~25% H₂O ≥98% (by iodometry) USP, BP, Ph. Eur., pharma grade

Toimittaja: PanReac AppliChem
Vaara

APPC142357.1210EA 988 EUR
APPC142357.1210
Bentsoyyliperoksidi, humidified with ~25% H₂O ≥98% (by iodometry) USP, BP, Ph. Eur., pharma grade
Bentsoyyliperoksidi
Kaava: C₁₄H₁₀O₄
MW: 242,23 g/mol
Sulamispiste: 105 °C
Storage Temperature: Jääkaappi
MDL numero: MFCD00003071
CAS-numero: 94-36-0
EINECS: 202-327-6
UN: 3104
ADR: 5.2,

Order Now

Testitulosten erittely

Assay (Iodometry) 70.0 - 77.0%
Minimum Assay (Iodometry) 98%
Identity According to Pharmacopoeias Passes Test
Acidity Passes Test
Chloride (Cl) 0.4%
Residual Solvents (Ph. Eur./USP) Passes Test
Benzaldehyde 0.25%
Unspecified Impurities 0.1%
Benzoic Acid 1.5%
Ethyl Benzoate 0.25%
Individual Impurity 1.5%
Total Impurities 2.0%
Water (H₂O) 20%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR