Strontiumnitraatti ≥99.0% (by complexometric analysis), Puriss. p.a. ACS, Fluka™
Toimittaja: Honeywell Chemicals
Vaara
31633-50KGEA 4240.02 EUR
HONE31633.9050 31633-50KG
Strontiumnitraatti ≥99.0% (by complexometric analysis), Puriss. p.a. ACS, Fluka™
Strontiumnitraatti
Kaava: Sr(NO₃)₂
MW: 211,63 g/mol
Sulamispiste: 645 °C
Tiheys: 2,980 g/cm3 (20 °C)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00011248
CAS-numero: 10042-76-9
EINECS: 233-131-9
UN: 1507
ADR: 5.1,III
Merck Index: 12,09007
Order Now
Testitulosten erittely
Assay (complexometric) Min. 99.0 %
Loss on drying (105°C, 4 h) Max. 0.1 %
pH (5 %, 25 °C) 5.0 - 7.0
In H₂O insoluble matter (10% solution) Max. 0.01 %
Ammonium (NH₄) Max. 0.005 %
Barium (Ba) Max. 0.01 %
Calcium (Ca) Max. 0.005 %
Copper (Cu) Max. 0.0002 %
Iron (Fe) Max. 0.0005 %
Potassium (K) Max. 0.005 %
Magnesium (Mg) Max. 0.002 %
Sodium (Na) Max. 0.005 %
Lead (Pb) Max. 0.0002 %
Zinc (Zn) Max. 0.0002 %
Heavy metals (as Pb) Max. 0.0005 %
Chloride (Cl) Max. 0.001 %
Sulfate (SO₄) Max. 0.005 %
Mg and alkali salts (SO₄) Max. 0.15 %
Appearance complying