UV/vis spectrophotometer, 7415
Toimittaja: Cole Parmer
634-1151EA 6270 EUR
634-1151
UV/vis spectrophotometer, 7415
Spektrofotometrit ja spektrometrit UV/Vis-spektrofotometrit
This UV/vis spectrophotometer offers 5 nm spectral bandwidth, simple photometrics, concentration, multi-wavelength, spectrum scanning, kinetics, to monitor the change in absorbance over time.
  • Internal memory for methods and results with 10 GB onboard storage
  • Multiple USB ports for data storage and printer connectivity
  • Multi-language options including English, French, German, Spanish and Italian
  • Features android operating system
For teaching, routine sample analysis in quality control and industrial applications.
The large 7" high definition colour touchscreen enables users to zoom in and out and select points all by simply touching the screen. The touchscreen interface is fast and responsive even when wearing gloves.
Toimitustiedot: Supplied with a 10×10 mm cuvette holder in sample and reference position, instructions manual and power supply with UK, EU and US power leads.