VWR® WL , Differential agar
Toimittaja: VWR
VWR®
101215AAEA 68 EUR
101215AA
VWR® WL , Differential agar
Elatusaineet mikrobiologiaan Elatusaineet mikrobiologiaan, jauhemaiset
WL differential agar is primarily for use in the isolation of bacteria encountered in brewing and industrial fermentation processes.