Dietyylieetteri, kuivattu (enint. 0,0075% H2O) stabiloitu ACS, ISO

Synonyms: Eetteri

Stabilization: Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol) ~6 ppm

APPC482770.0311EA 141 EUR
APPC482770.0311
Dietyylieetteri, kuivattu (enint. 0,0075% H2O) stabiloitu ACS, ISO
Dietyylieetteri
Kaava: (CH₃CH₂)₂O
MW: 74,12 g/mol
Kiehumispiste: 34,6 °C
Sulamispiste: –116 °C
Tiheys: 0,715 kg/l
MDL numero: MFCD00011646
CAS-numero: 60-29-7
EINECS: 200-467-2
UN: 1155
ADR: 3,I
https://fi.vwr.com/stibo/web/std.lang.all/02/58/730258.jpg

Order Now

Testitulosten erittely

Minimum assay (G.C.) 99,8%
Identity IR passes test
Density at 20/4 0,713 - 0,715
APHA colour 10
Acidity ≤0,0002 meq/g
Non-volatile matter ≤0,001%
Darkened substances by H₂SO₄ passes test
Carbonyl compounds (as HCHO) ≤0,001%
Peroxides (as H₂O₂) ≤0,00003%
Acetone (GC) ≤0,005%
Methanol (GC) ≤0,02%
Ethanol (GC) ≤0,05%
Water (H₂O) ≤0,0075%
Metals by ICP [in mg/Kg (ppm)], Ag 0,05
Metals by ICP [in mg/Kg (ppm)], Al 0,5
Metals by ICP [in mg/Kg (ppm)], As 0,05
Metals by ICP [in mg/Kg (ppm)], Au 0,05
Metals by ICP [in mg/Kg (ppm)], B 0,02
Metals by ICP [in mg/Kg (ppm)], Ba 0,1
Metals by ICP [in mg/Kg (ppm)], Be 0,02
Metals by ICP [in mg/Kg (ppm)], Bi 0,05
Metals by ICP [in mg/Kg (ppm)], Ca 0,5
Metals by ICP [in mg/Kg (ppm)], Cd 0,05
Metals by ICP [in mg/Kg (ppm)], Co 0,02
Metals by ICP [in mg/Kg (ppm)], Cr 0,02
Metals by ICP [in mg/Kg (ppm)], Cu 0,02
Metals by ICP [in mg/Kg (ppm)], Fe 0,1
Metals by ICP [in mg/Kg (ppm)], Ga 0,02
Metals by ICP [in mg/Kg (ppm)], Ge 0,05
Metals by ICP [in mg/Kg (ppm)], Hg 0,05
Metals by ICP [in mg/Kg (ppm)], In 0,05
Metals by ICP [in mg/Kg (ppm)], K 0,1
Metals by ICP [in mg/Kg (ppm)], Li 0,05
Metals by ICP [in mg/Kg (ppm)], Mg 0,1
Metals by ICP [in mg/Kg (ppm)], Mn 0,02
Metals by ICP [in mg/Kg (ppm)], Mo 0,02
Metals by ICP [in mg/Kg (ppm)], Na 0,5
Metals by ICP [in mg/Kg (ppm)], Ni 0,02
Metals by ICP [in mg/Kg (ppm)], P 0,2
Metals by ICP [in mg/Kg (ppm)], Pb 0,1
Metals by ICP [in mg/Kg (ppm)], Pt 0,02
Metals by ICP [in mg/Kg (ppm)], S 0,2
Metals by ICP [in mg/Kg (ppm)], Sb 0,02
Metals by ICP [in mg/Kg (ppm)], Si 0,2
Metals by ICP [in mg/Kg (ppm)], Sn 0,1
Metals by ICP [in mg/Kg (ppm)], Sr 0,2
Metals by ICP [in mg/Kg (ppm)], Ti 0,02
Metals by ICP [in mg/Kg (ppm)], Tl 0,02
Metals by ICP [in mg/Kg (ppm)], V 0,02
Metals by ICP [in mg/Kg (ppm)], Zn 0,1
Metals by ICP [in mg/Kg (ppm)], Zr 0,02

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR