Alkaline earth metal mix for ICP, 5 elements, TraceCERT®, Supelco®
Toimittaja: Merck
SUPL67288-100MLEA 108 EUR
SUPL67288-100ML
Alkaline earth metal mix for ICP, 5 elements, TraceCERT®, Supelco®
100 mg/L each: Be, Mg, Ca, Sr and Ba in 2% nitric acid
Sertifikaatit: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034 (ISO Guide 34).