DREL complete water quality lab
Toimittaja: Hach
HACH2922401EA 7590 EUR
HACH2922401
DREL complete water quality lab
Ympäristöanalytiikan pikatestit
This complete water lab contains a DR 1900 portable spectrophotometer and digital titrator, along with reagents and apparatus necessary to run approximately 100 tests on 24 different parameters in a rugged, well-organized carrying case.
  • Contains durable instruments, premeasured reagents impervious to environmental contamination, and apparatus and labware
  • Instruments have the option of running on batteries, so tests can be run without concern for sample preservation and handling
  • Rugged kit is designed to withstand harsh field conditions
  • All-new DR1900 portable spectrophotometer is incredibly light (1,49 kg) with battery, and can read Hach TNT+ vials
Two test kit cases with molded inserts are included. One case holds instrument and accessories, the other holds reagents and testing apparatus. The instrument case has additional storage space for adding optional pH meter, portable turbimeter, and two probes.
Toimitustiedot: Each laboratory contains the following: DR 1900 Spectrophotometer, Rechargeable Batter, 1" Matched Glass Sample Cells, Instrument Case, Reagent/Apparatus Case, Reagent Set, Apparatus Set, Cell Adapters, Power Supply.