Ashing furnaces with fold down door, Tmax 1100 °C and optional controller C450, LV 3/11 - LV 15/11
Toimittaja: Nabertherm
NABELV094K1ON1EA 0 EUR
NABELV094K1ON1 NABELV094K1ON1TZ NABELV034K1ON1 NABELV094K1ON1Z NABELV034K1ON1TZ NABELV154K1ON1T NABELV154K1ON1Z NABELV054K1ON1TZ NABELV054K1ON1Z NABELV034K1ON1Z NABELV154K1ON1 NABELV054K1ON1 NABELV154K1ON1TZ
Ashing furnaces with fold down door, Tmax 1100 °C and optional controller C450, LV 3/11 - LV 15/11
The LV 3/11 to LV 15/11 ashing furnaces are specially designed for ashing in the laboratory. A special air intake and exhaust system allows air exchange greater than six times per minute. Incoming air is preheated to ensure good temperature uniformity.
  • Heating from two sides
  • Ceramic heating plates with integral heating element which is protected from fumes and splashing, and easy to replace
  • Exclusive use of insulation materials without categorization according to EC Regulation No 1272/2008 (CLP). This explicitly means that alumino silicate wool, also known as “refractory ceramic fibre” (RCF), which is classified and possibly carcinogenic, is not used
  • Fold down door (LV) which can be used as work platform
The furnace is equipped with the C 550 controller with LC display, featuring 10 freely adjustable programs and 20 segments and 2 extra functions. These controllers are PID microprocessor controlled with a self-diagnostic system and integrated process documentation.
Minutes to Tmax without load and standard voltage: 120
Tilaustiedot: Optional accessories such as over-temperature limit controller with adjustable temperature cut out for thermal protection Class 2, in accordance with EN 60519-2 as temperature limiter to protect the oven and load, access port for thermocouple.
Varoitus: Maximum continuous operating temperature should be 50 °C below the 'Max. temp.' of the model shown in the table above.
Outer dimensions include exhaust tube (Ø 80 mm).