Kalsiumfluoridi 99.0-102.0%, Puriss., meets analytical specification of DAB, Fluka™
Synonyms:
Fluorite
, Fluorspar
HONE01123.9040
Kalsiumfluoridi 99.0-102.0%, Puriss., meets analytical specification of DAB, Fluka™
Kalsiumfluoridi
Kaava:
CaF₂ MW: 78,07 g/mol Kiehumispiste: 2,500 °C Sulamispiste: 1,423 °C Tiheys: 3,18 g/cm3 Storage Temperature: Huoneenlämpö |
MDL numero:
MFCD00010907 CAS-numero: 7789-75-5 EINECS: 232-188-7 |
Testitulosten erittely
Assay | 99.0 - 102.0 % |
Loss on drying (105°C, 2 h) | Max. 0.5 % |
Free acid (as HF) | Max. 0.1 % |
Arsenic (As) | Max. 0.0005 % |
Iron (Fe) | Max. 0.01 % |
Lead (Pb) | Max. 0.002 % |
Heavy metals (as Pb) | Max. 0.005 % |
Chloride (Cl) | Max. 0.1 % |
Sulfate (SO₄) | Max. 0.005 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...