Peristaltic pump, multi channel, PLP380
Toimittaja: Behr
BEHRB00454739EA 0 EUR
BEHRB00454739
Peristaltic pump, multi channel, PLP380
Pumput Peristalttiset pumput
High flexibility of chemical resistance.
  • High speed range
  • Controlling through external signals is possible
  • Controlling through a computer is possible
For the Dülabo peristaltic pump PLP380 there are different pump heads which can be applied, this results in flexibility in your day to day usage, enabling multiple tubing sizes being applicable. Depending on the tubing material in use, this pump type can be used for various chemicals, korro-sive and aggressive as well as sterile solutions. The user friendly membrane keyboard enables the user to regulate the flow speed (RPM) using the switch arrays. Furthermore, the user can make use of the special button on the membrane keyboard which activates a "quick flow mode“. Via RS485 or other external adapters for other signals, steering the pump externally is also possible. Additionally a foot switch can be attached to the pump to enable the pumping function as long as the foot switch is activated.
Tilaustiedot: Various pump heads and accessories are available and must be ordered separately.