Dialyser floating racks, DiaEasy™
Toimittaja: BioVision
BIOV1000-10EA 113 EUR
BIOV1000-10 BIOV1020-5 BIOV1018-10 BIOV1016-10 BIOV1016-5 BIOV1020-10 BIOV1000-5 BIOV1018-5
Dialyser floating racks, DiaEasy™
Balance dialysis tubes at two different heights for buffer exchange during dialysis.
  • Convenient, sturdy floating racks to balance dialysis tubes for buffer exchange during dialysis
  • Each of these racks can hold up to four tubes
These racks are designed to support DiaEasy™ dialyser tubes of different volume.