APLIA7626
Toimittaja: PanReac AppliChem
A7626.00005EA 196 EUR
A7626.00005
APLIA7626
Staurosporine was isolated from Streptomyces staurosporeus. It has antifungal but not antibacterial properties.
Staurosporine is cell-permeable and inhibits several kinases at very low concentrations.