Pesticide reference standard, Pyrifenox, PESTANAL®, analytical standard, Supelco®
Toimittaja: Merck
PESTANAL®
45737-100MGEA 100 EUR
45737-100MG
Pesticide reference standard, Pyrifenox, PESTANAL®, analytical standard, Supelco®
Standardiaineet ja vertailumateriaalit Organic Standards
Pyrifenox may be used as a reference standard for the determination of pyrifenox in samples by modified QuEChERS and ultra-high performance liquid chromatography coupled to tandem mass spectrometry (UHPLC–MS/MS).
  • Used in controlling powdery mildews of field-grown fruits, mainly wine grapes, mango and nectarine trees