Organic reference standard, Glutaric semialdehyde, ≥85,0% (GC), Supelco®
Toimittaja: Merck
SUPL00380-50MGEA 822 EUR
SUPL00380-50MG SUPL00380-10MG
Organic reference standard, Glutaric semialdehyde, ≥85,0% (GC), Supelco®
Metabolite of lysine degradation. In the lysine degradation IV pathway, glutarate semialdehyde reacts with NADP⁺ and H₂O to produce glutarate, NADPH, and H⁺. In this pathway, glutarate semialdehyde is produced by the reaction between 5-aminopentanoate and 2-ketoglutarate, with L-glutamate as a byproduct.