Rikkihappo 40% for determination of gas metabolism acc. to Knipping

Vaara

86171.320EA 83.1 EUR
86171.320
Rikkihappo 40% for determination of gas metabolism acc. to Knipping
Rikkihappo
Kaava: H₂SO₄
MW: 98,08 g/mol
MDL numero: MFCD00064589
CAS-numero: 7664-93-9
UN: 2796
ADR: 8,II

Order Now

Testitulosten erittely

Assay (m/m) 39.60 – 40.40 %
Appearance complies
Residue after ignition max. 50 ppm
Substances reducing potassium permanganate (as SO₂) max. 10 ppm
Chloride (Cl) max. 2 ppm
Nitrate (NO₃) max. 2 ppm
Heavy metals (as Pb) max. 5 ppm
Ammonium (NH4) max. 5 ppm
Arsenic (As) max. 0,1 ppm
Iron (Fe) max. 2 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR