Selenium Standard for ICP-MS, TraceCERT®, Supelco®
Toimittaja: Merck
SUPL56544-100MLEA 89.7 EUR
SUPL56544-100ML
Selenium Standard for ICP-MS, TraceCERT®, Supelco®
Sertifikaatit: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034 (ISO Guide 34)