Headspace wash kit for PAL autosamplers, VWR®
Toimittaja: VWR, part of Avantor
VWR®
548-2363EA 76.2 EUR
548-2363
Headspace wash kit for PAL autosamplers, VWR®
Kromatografiaan tarvikkeet Kromatografiaan varaosat
This wash kit is a convenient all in one solution for PAL autosampler.
  • Improved sealing, less evaporation or contamination of wash solvents
  • Easy to apply caps
  • Reproducible syringe clean up