Splash shower tester
Toimittaja: Bradley
118-0798EA 589 EUR
BRLY20-S19-330ST
Splash shower tester
Plastic collection bag with handle to easily collect the water in a rain shower. Useful for flushing the shower and checking the water yield.