Etikkahappo ≥100%, LiChropur™ HPLC-analytiikkaan, Supelco®
Toimittaja: Merck
Vaara
M-Clarity™ quality level = MQ100
5.43808.0100EA 60.4 EUR
5.43808.0100 5.43808.0250
Etikkahappo ≥100%, LiChropur™ HPLC-analytiikkaan, Supelco®
Etikkahappo
Kaava: H₃CCOOH
MW: 60,05 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00036152
CAS-numero: 64-19-7
EINECS: 200-580-7
UN: 2789
ADR: 8 (3),II
https://fi.vwr.com/stibo/web/std.lang.all/31/15/723115.jpg
For metal trace analysis in AAS or ICP
Ask for your “customized multi-element standard solution” with your exact composition using the request form
Ask for a quotation
12/31/2022
Order Now
For metal trace analysis in AAS or ICP
Ask for your “customized multi-element standard solution” with your exact composition using the request form
Ask for a quotation
12/31/2022