Signs, COVID-19 reception, PP, Brady®, W.H.Brady
Toimittaja: Brady
BRDY306931EA 29.9 EUR
BRDY306931 BRDY306923 BRDY306933
Signs, COVID-19 reception, PP, Brady®, W.H.Brady
Merkit, kyltit ja lattiamerkinnät
Signs are made of surface printed rigid polypropylene material with a slightly textured surface. They are overlaminated with a gloss clear polyester.
  • Ideal for indoor and outdoor applications
  • Well suited for harsh environments
  • Good solvent resistance
  • Rectangular shape
  • Colours: Black and white on red, green, blue, yellow and white
Thickness: 1,501 mm.