Streptavidin, Cy2®
Toimittaja: Cytiva
PA42001EA 427 EUR
GEHEPA42001
Streptavidin, Cy2®
Streptavidin-fluor conjugates suitable for use in the fluorescent analysis of biotinylated samples.
  • Streptavidin conjugated to a range of fluorescent dyes for use in in situ hybridisation and flow cytometry
  • High specificity reagents