D-(+)-Cystine

Toimittaja: Molekula

MOLE14591553-1GEA 0 EUR
MOLE14591553-1G
D-(+)-Cystine
D-(+)-Cystine
Kaava: HOOCCH(NH₂)CH₂SSCH₂CH(NH₂)COOH
MW: 240,3 g/mol
Sulamispiste: 256…261 °C
MDL numero: MFCD00002610
CAS-numero: 349-46-2
EINECS: 206-486-2
Merck Index: 13,02811

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR