4-Amino-2,6-dimethoxypyrimidine ≥98.0% (by GC, titration analysis)

Toimittaja: TCI EUROPE
Varoitus

TCIAA2264-5GEA 41.8 EUR
TCIAA2264-5G
4-Amino-2,6-dimethoxypyrimidine ≥98.0% (by GC, titration analysis)
4-Amino-2,6-dimethoxypyrimidine
Kaava: C₆H₉N₃O₂
MW: 155,16 g/mol
MDL numero: MFCD00082802
CAS-numero: 3289-50-7
EINECS: 221-946-2

Order Now

Testitulosten erittely

Appearance White to Light yellow to Green powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 149.0 to 153.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR