5-Aminopyrimidine ≥98.0% (by GC, titration analysis)

Toimittaja: TCI EUROPE
Varoitus

TCIAA2589-1GEA 86.9 EUR
TCIAA2589-1G
5-Aminopyrimidine ≥98.0% (by GC, titration analysis)
5-Aminopyrimidine
Kaava: C₄H₅N₃
MW: 95,1000 g/mol
Sulamispiste: 171°C
MDL numero: MFCD01529870
CAS-numero: 591-55-9

Order Now

Testitulosten erittely

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR