1-Boc-(2-aminomethyl)piperidine ≥98.0% (by titrimetric analysis)

Toimittaja: TCI EUROPE
Varoitus

TCIAA2713-1GEA 83.6 EUR
TCIAA2713-1G
1-Boc-(2-aminomethyl)piperidine ≥98.0% (by titrimetric analysis)
1-Boc-(2-aminomethyl)piperidine
Kaava: C₁₁H₂₂N₂O₂
MW: 214,31 g/mol
Leimahduspiste: 110°C
MDL numero: MFCD01317791
CAS-numero: 370069-31-1

Order Now

Testitulosten erittely

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 95.0 %
Purity (Nonaqueous Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR