4'-Acetamidoacetophenone ≥98.0% (by GC)
Toimittaja: TCI EUROPE
|
Varoitus
|
TCIAA2821-5GEA
52.8
EUR
TCIAA2821-5G
TCIAA2821-25G
4'-Acetamidoacetophenone ≥98.0% (by GC)
4'-Acetamidoacetophenone
Kaava:
C₁₀H₁₁NO₂ MW: 177,20 g/mol |
MDL numero:
MFCD00014965 CAS-numero: 2719-21-3 EINECS: 220-320-6 |
Testitulosten erittely
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 169.0 to 173.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...