4-(Bromomethyl)benzophenone ≥96.0% (by GC)

Toimittaja: TCI EUROPE
Varoitus

TCIAB3840-5GEA 132 EUR
TCIAB3840-5G
4-(Bromomethyl)benzophenone ≥96.0% (by GC)
4-(Bromomethyl)benzophenone
Kaava: C₁₄H₁₁BrO
MDL numero: MFCD00092076
CAS-numero: 32752-54-8
EINECS: 251-197-7

Order Now

Testitulosten erittely

Appearance Light orange to Yellow to Green powder to crystal
Purity (GC) min. 96.0 %
Melting point 110.0 to 114.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR