3-Bromophenylsulphur pentafluoride ≥92.0% (by GC)
Toimittaja: TCI EUROPE
|
Varoitus
|
TCIAB3866-1GEA
168
EUR
TCIAB3866-1G
3-Bromophenylsulphur pentafluoride ≥92.0% (by GC)
3-Bromophenylsulphur pentafluoride
Kaava:
C₆H₄BrF₅S MW: 283,06 g/mol Kiehumispiste: 82°C Leimahduspiste: 88°C |
MDL numero:
MFCD03425927 CAS-numero: 672-30-0 |
Testitulosten erittely
Appearance | Colorless to Light yellow to Red clear liquid |
Purity (GC) | min. 92.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...