2-Bromoquinoxaline ≥98.0% (by GC)

Toimittaja: TCI EUROPE
Varoitus

TCIAB4632-1GEA 64.9 EUR
TCIAB4632-1G TCIAB4632-200MG
2-Bromoquinoxaline ≥98.0% (by GC)
2-Bromoquinoxaline
Kaava: C₈H₅BrN₂
MW: 209,04 g/mol
MDL numero: MFCD07368206
CAS-numero: 36856-91-4

Order Now

Testitulosten erittely

Appearance White to Light red to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 55.0 to 59.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR