3-Bromoimidazo[1,2-b]pyridazine ≥98.0% (by GC, titration analysis)

Toimittaja: TCI EUROPE

TCIAB4652-1GEA 148 EUR
TCIAB4652-1G
3-Bromoimidazo[1,2-b]pyridazine ≥98.0% (by GC, titration analysis)
3-Bromoimidazo[1,2-b]pyridazine
Kaava: C6H4BrN3
MW: 198,0201 g/mol
MDL numero: MFCD09757672
CAS-numero: 18087-73-5

Order Now

Testitulosten erittely

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 148.0 to 152.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR