2-Bromo-13,13-dimethyl-6H-indeno[1,2-b]anthracene-6,11(13H)-dione ≥98.0% (by GC)

Toimittaja: TCI EUROPE

TCIAB4668-1GEA 99 EUR
TCIAB4668-1G
2-Bromo-13,13-dimethyl-6H-indeno[1,2-b]anthracene-6,11(13H)-dione ≥98.0% (by GC)
2-Bromo-13,13-dimethyl-6H-indeno[1,2-b]anthracene-6,11(13H)-dione
Kaava: C₂₃H₁₅BrO₂
MW: 403,28 g/mol
MDL numero: MFCD20275103
CAS-numero: 1196107-73-9

Order Now

Testitulosten erittely

Appearance Light yellow to Yellow to Orange powder to crystal
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR