2-Bromo-4-fluoro-6-nitroaniline ≥98.0% (by GC)

Toimittaja: TCI EUROPE
Varoitus

TCIAB4872-1GEA 121 EUR
TCIAB4872-1G
2-Bromo-4-fluoro-6-nitroaniline ≥98.0% (by GC)
2-Bromo-4-fluoro-6-nitroaniline
Kaava: C₆H₄BrFN₂O₂
MW: 235,01 g/mol
MDL numero: MFCD07779528
CAS-numero: 10472-88-5

Order Now

Testitulosten erittely

Appearance Light yellow to Brown powder to crystal
Purity (GC) min. 98.0 %
Melting point 75.0 to 79.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR