(2,2'-Bipyridine)bis[2-(2,4-difluorophenyl)pyridine]iridium(III) Hexafluorophosphate ≥80.0% (by HPLC)

Toimittaja: TCI EUROPE
Varoitus

TCIAB4944-200MGEA 205 EUR
TCIAB4944-200MG
(2,2'-Bipyridine)bis[2-(2,4-difluorophenyl)pyridine]iridium(III) Hexafluorophosphate ≥80.0% (by HPLC)
(2,2'-Bipyridine)bis[2-(2,4-difluorophenyl)pyridine]iridium(III) Hexafluorophosphate
Kaava: C₃₂H₂₀F₁₀IrN₄P
MW: 873,71 g/mol
MDL numero: MFCD29472513
CAS-numero: 864163-80-4

Order Now

Testitulosten erittely

Appearance Light yellow to Brown powder to crystal
Purity (HPLC) min. 80.0 area%
Elemental analysis (Nitrogen) 6.00 to 6.75 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR