2'-Bromo-4-chlorobiphenyl ≥98.0% (by GC)

Toimittaja: TCI EUROPE

TCIAB5253-1GEA 75.9 EUR
TCIAB5253-1G
2'-Bromo-4-chlorobiphenyl ≥98.0% (by GC)
2'-Bromo-4-chlorobiphenyl
Kaava: C₁₂H₈BrCl
MW: 267,55 g/mol
MDL numero: MFCD18072809
CAS-numero: 179526-95-5
UN: 3152
ADR: 9,II

Order Now

Testitulosten erittely

Appearance White to Orange to Green powder to lump
Purity (GC) min. 98.0 %
Melting point 39.0 to 43.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR