(1R,8S,9s)-Bicyclo[6.1.0]non-4-yn-9-ylmethanol ≥90.0% (by GC)

Toimittaja: TCI EUROPE

TCIAB5467-100MGEA 300 EUR
TCIAB5467-100MG
(1R,8S,9s)-Bicyclo[6.1.0]non-4-yn-9-ylmethanol ≥90.0% (by GC)
(1R,8S,9s)-Bicyclo[6.1.0]non-4-yn-9-ylmethanol
Kaava: C₁₀H₁₄O
MW: 150,22 g/mol
Storage Temperature: Pakastin
MDL numero: MFCD26142970
CAS-numero: 1263166-90-0

Order Now

Testitulosten erittely

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 90.0 %
Elemental analysis (Carbon) 76.00 to 84.00 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR