10-Bromo-7-phenyl-7H-benzo[c]carbazole ≥98.0% (by HPLC, total nitrogen)

Toimittaja: TCI EUROPE
Varoitus

TCIAB5868-1GEA 224 EUR
TCIAB5868-1G
10-Bromo-7-phenyl-7H-benzo[c]carbazole ≥98.0% (by HPLC, total nitrogen)
10-Bromo-7-phenyl-7H-benzo[c]carbazole
Kaava: C₂₂H₁₄BrN
MW: 372,27 g/mol
MDL numero: MFCD26390974
CAS-numero: 1210469-11-6

Order Now

Testitulosten erittely

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR