(1R)-(+)-2,10,Camphorsultam ≥98.0% (by GC)

Toimittaja: TCI EUROPE
Varoitus

TCIAC1324-5GEA 322 EUR
TCIAC1324-5G
(1R)-(+)-2,10,Camphorsultam ≥98.0% (by GC)
(1R)-(+)-2,10,Camphorsultam
Kaava: C₁₀H₁₇NO₂S
MW: 215,32 g/mol
MDL numero: MFCD00151762
CAS-numero: 108448-77-7

Order Now

Testitulosten erittely

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 182.0 to 186.0 deg-C
Specific rotation [a]20/D +30.0 to +35.0 deg (C=1, CHCl3)
Solubility in Chloroform almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR