Carbon nanotube multi-walled, Length: 1-2 µm, Ø 40-60 nm

Toimittaja: TCI EUROPE
Varoitus

Synonyms: CNT

TCIAC2157-1GEA 97.9 EUR
TCIAC2157-1G
Carbon nanotube multi-walled, Length: 1-2 µm, Ø 40-60 nm
Carbon nanotube
Kaava: C
MW: 12,011 g/mol
MDL numero: MFCD00133992
CAS-numero: 308068-56-6

Order Now

Testitulosten erittely

Appearance Black powder to crystal
Purity (TPO Method) min. 95 %
Diameter 40 to 60 nm
Length 1 to 2 micro_m

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR