4-Cyano-3-fluorobenzoic acid ≥97.0% (by HPLC, titration analysis)
Toimittaja: TCI EUROPE
|
Varoitus
|
TCIAC2799-1GEA
160
EUR
TCIAC2799-1G
4-Cyano-3-fluorobenzoic acid ≥97.0% (by HPLC, titration analysis)
4-Cyano-3-fluorobenzoic acid
Kaava:
C₈H₄FNO₂ MW: 165,1213 g/mol |
MDL numero:
MFCD03094324 CAS-numero: 176508-81-9 |
Testitulosten erittely
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 97.0 area% |
Purity (Neutralization titration) | min. 97.0 % |
Melting point | 162.0 to 166.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...