3-Cyanochromone ≥98.0% (by GC)

Toimittaja: TCI EUROPE
Vaara

TCIAC3075-5GEA 70.4 EUR
TCIAC3075-5G
3-Cyanochromone ≥98.0% (by GC)
3-Cyanochromone
Kaava: C₁₀H₅NO₂
MW: 171,16
MDL numero: MFCD00052604
CAS-numero: 50743-17-4
UN: 3439
ADR: 6.1,III

Order Now

Testitulosten erittely

Appearance White to Yellow to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 175.0 to 179.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR