3-Cyclohexylthiophene ≥96.0% (by GC)
Toimittaja: TCI EUROPE
TCIAC3400-5GEA
381
EUR
TCIAC3400-5G
TCIAC3400-1G
3-Cyclohexylthiophene ≥96.0% (by GC)
3-Cyclohexylthiophene
Kaava:
C₁₀H₁₄S MW: 166,28 g/mol Storage Temperature: Jääkaappi |
MDL numero:
MFCD02260242 CAS-numero: 120659-34-9 |
Testitulosten erittely
Appearance | White to Almost white clear liquid |
Purity (GC) | min. 96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...