5-Decanol ≥96.0% (by GC)

Toimittaja: TCI EUROPE
Varoitus

TCIAD1381-5MLEA 169 EUR
TCIAD1381-5ML
5-Decanol ≥96.0% (by GC)
5-Decanol
Kaava: C₁₀H₂₂O
Kiehumispiste: 80°C
Sulamispiste: 9°C
MDL numero: MFCD00039626
CAS-numero: 5205-34-5
EINECS: 225-999-2

Order Now

Testitulosten erittely

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR