Diisopropyl disulphide ≥98.0% (by GC)

Toimittaja: TCI EUROPE
Vaara

TCIAD2582-25GEA 50.6 EUR
TCIAD2582-25G
Diisopropyl disulphide ≥98.0% (by GC)
Diisopropyl disulphide
Kaava: C₆H₁₄S₂
MW: 150,31 g/mol
Kiehumispiste: 177°C
Leimahduspiste: 18°C
MDL numero: MFCD00008894
CAS-numero: 4253-89-8
EINECS: 224-225-0
UN: 1993
ADR: 3,II

Order Now

Testitulosten erittely

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR