Bis(propylcyclopentadienyl)hafnium dichloride ≥98.0% (by titrimetric analysis)

Toimittaja: TCI EUROPE
Varoitus

TCIAD3286-1GEA 212 EUR
TCIAD3286-1G
Bis(propylcyclopentadienyl)hafnium dichloride ≥98.0% (by titrimetric analysis)
Bis(propylcyclopentadienyl)hafnium dichloride
Kaava: C₁₆H₂₂Cl₂Hf
CAS-numero: 85722-06-1

Order Now

Testitulosten erittely

Appearance White to Gray to Brown powder to crystal
Purity (Chelometric Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR