1,3-Dimethyl-1H-pyrazole ≥98.0% (by GC)
Toimittaja: TCI EUROPE
|
Varoitus
|
TCIAD4528-5GEA
55
EUR
TCIAD4528-5G
1,3-Dimethyl-1H-pyrazole ≥98.0% (by GC)
1,3-Dimethyl-1H-pyrazole
Kaava:
C₅H₈N₂ MW: 96,1304 g/mol Kiehumispiste: 138°C |
MDL numero:
MFCD00036665 CAS-numero: 694-48-4 UN: 1993 ADR: 3,III |
Testitulosten erittely
Appearance | Light orange to Yellow to Green clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...