2,3-Dihydrobenzofuran-5-boronic acid (contains varying amounts of Anhydride)

Toimittaja: TCI EUROPE
Varoitus

TCIAD4711-200MGEA 114 EUR
TCIAD4711-200MG
2,3-Dihydrobenzofuran-5-boronic acid (contains varying amounts of Anhydride)
2,3-Dihydrobenzofuran-5-boronic acid
Kaava: C₈H₉BO₃
MW: 163,97 g/mol
Sulamispiste: 240°C
MDL numero: MFCD02681979
CAS-numero: 227305-69-3

Order Now

Testitulosten erittely

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) 97.0 to 112.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR