9,10-Dihydroacridine ≥98.0% (by GC)
Toimittaja: TCI EUROPE
Varoitus
TCIAD5058-1GEA 50.6 EUR
TCIAD5058-1G TCIAD5058-5G
9,10-Dihydroacridine ≥98.0% (by GC)
9,10-Dihydroacridine
Kaava: C₁₃H₁₁N
MW: 181,24 g/mol
Kiehumispiste: 348°C
MDL numero: MFCD00022256
CAS-numero: 92-81-9
EINECS: 202-192-3
Order Now
Testitulosten erittely
Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 168.0 to 172.0 deg-C