2,6-Diphenylanthracene ≥98.0% (by GC), purified by sublimation

Toimittaja: TCI EUROPE

TCIAD5152-100MGEA 240 EUR
TCIAD5152-100MG
2,6-Diphenylanthracene ≥98.0% (by GC), purified by sublimation
2,6-Diphenylanthracene
Kaava: C₂₆H₁₈
MW: 330,43 g/mol
CAS-numero: 95950-70-2

Order Now

Testitulosten erittely

Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR