exo-DNTT-PMI (DNTT-Precursor) ≥95.0% (by HPLC)
Toimittaja: TCI EUROPE
TCIAD5154-50MGEA
791
EUR
TCIAD5154-50MG
exo-DNTT-PMI (DNTT-Precursor) ≥95.0% (by HPLC)
exo-DNTT-PMI (DNTT-Precursor)
Kaava:
C₃₂H₁₉NO₂S₂ MW: 513,63 g/mol |
CAS-numero:
1269669-43-3 |
Testitulosten erittely
Appearance | White to Light yellow to Dark green powder to crystal |
Purity (HPLC) | min. 95.0 area% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...