1,2-Dierucoyl-sn-glycero-3-phosphocholine ≥98.0% (by titrimetric analysis)
Toimittaja: TCI EUROPE
TCIAD5319-250MGEA
182
EUR
TCIAD5319-250MG
1,2-Dierucoyl-sn-glycero-3-phosphocholine ≥98.0% (by titrimetric analysis)
1,2-Dierucoyl-sn-glycero-3-phosphocholine
Kaava:
C₅₂H₁₀₀NO₈P MW: 898,34 g/mol Storage Temperature: Pakastin |
MDL numero:
MFCD00674315 CAS-numero: 51779-95-4 |
Testitulosten erittely
Appearance | White to Almost white powder to crystal |
Purity (HPLC) (CAD) | min. 95.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...